Product Details

SNP ID
rs3810421
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3365641 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCATACCTCAGTCTTGAGATCTG[C/T]GAAAAGGGCACACCCCTGAGGCAGG
Phenotype
MIM: 600729
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
NFIC PubMed Links
Additional Information
For this assay, SNP(s) [rs116381300] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
NFIC
Gene Name
nuclear factor I C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001245002.1 Intron NP_001231931.1
NM_001245004.1 Intron NP_001231933.1
NM_001245005.1 Intron NP_001231934.1
NM_005597.3 Intron NP_005588.2
NM_205843.2 Intron NP_995315.1
XM_005259564.3 Intron XP_005259621.1
XM_006722759.2 Intron XP_006722822.2
XM_017026834.1 Intron XP_016882323.1
XM_017026835.1 Intron XP_016882324.1
XM_017026836.1 Intron XP_016882325.1

View Full Product Details