Product Details
- SNP ID
-
rs11878617
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:4769728 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GGACAGTTCCCAGCGGGCTCCGTTT[A/C]GTCTCCAGAACCCTGGACAGCTCCT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MIR7-3
PubMed Links
Gene Details
- Gene
- MIR7-3
- Gene Name
- microRNA 7-3
There are no transcripts associated with this gene.
- Gene
- MIR7-3HG
- Gene Name
- MIR7-3 host gene
There are no transcripts associated with this gene.
View Full Product Details