Product Details
- SNP ID
-
rs35070962
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:45149782 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAGGAGATGTTTATTGTCACATGT[A/G]GATGTTCAGAGTGGCAGGCAGCTCT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NKPD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76598479] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NKPD1
- Gene Name
- NTPase, KAP family P-loop domain containing 1
- Gene
- PPP1R37
- Gene Name
- protein phosphatase 1 regulatory subunit 37
There are no transcripts associated with this gene.
View Full Product Details