Product Details

SNP ID
rs5749081
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30369779 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCACACAGTGTGGCGGTTAAGAAC[A/G]CAGGCCTTGGGGATCATGCCTTGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC157 PubMed Links

Gene Details

Gene
CCDC157
Gene Name
coiled-coil domain containing 157
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017437.3 Intron NP_001017437.2
NM_001318334.1 Intron NP_001305263.1
NM_001318335.1 Intron NP_001305264.1
XM_006724278.3 Intron XP_006724341.1
XM_006724279.3 Intron XP_006724342.1
XM_006724280.3 Intron XP_006724343.1
XM_011530263.2 Intron XP_011528565.1
XM_011530266.2 Intron XP_011528568.1
XM_011530267.2 Intron XP_011528569.1
XM_011530268.2 Intron XP_011528570.1
XM_011530269.2 Intron XP_011528571.1
XM_011530270.2 Intron XP_011528572.1
Gene
KIAA1656
Gene Name
KIAA1656 protein
There are no transcripts associated with this gene.

Gene
RNF215
Gene Name
ring finger protein 215
There are no transcripts associated with this gene.

View Full Product Details