Product Details

SNP ID
rs758326
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:30235127 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGGTGGTGACCCCATGCCAGGCACT[A/G]CATTACACACCCCGGGTGCATTATC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF536 PubMed Links

Gene Details

Gene
ZNF536
Gene Name
zinc finger protein 536
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014717.2 Intron NP_055532.1
XM_011527554.2 Intron XP_011525856.1
XM_011527555.2 Intron XP_011525857.1
XM_011527557.2 Intron XP_011525859.1
XM_011527558.2 Intron XP_011525860.1
XM_011527560.2 Intron XP_011525862.1
XM_017027527.1 Intron XP_016883016.1
XM_017027528.1 Intron XP_016883017.1
XM_017027529.1 Intron XP_016883018.1
XM_017027530.1 Intron XP_016883019.1
XM_017027531.1 Intron XP_016883020.1
XM_017027532.1 Intron XP_016883021.1
XM_017027533.1 Intron XP_016883022.1
XM_017027534.1 Intron XP_016883023.1
XM_017027535.1 Intron XP_016883024.1
XM_017027536.1 Intron XP_016883025.1
XM_017027537.1 Intron XP_016883026.1
XM_017027538.1 Intron XP_016883027.1
XM_017027539.1 Intron XP_016883028.1
XM_017027540.1 Intron XP_016883029.1
XM_017027541.1 Intron XP_016883030.1
XM_017027542.1 Intron XP_016883031.1
XM_017027543.1 Intron XP_016883032.1

View Full Product Details