Product Details
- SNP ID
-
rs2072684
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:1985414 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GTAGAGCCGCCATTGTTGCTATAAG[C/G]CTGGTCAGTTAGAATGAAGTCTCTT
- Phenotype
-
MIM: 600924
MIM: 611256
MIM: 603927
MIM: 605915
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GFER
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74002752] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GFER
- Gene Name
- growth factor, augmenter of liver regeneration
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005262.2 |
|
Intron |
|
|
NP_005253.3 |
- Gene
- NOXO1
- Gene Name
- NADPH oxidase organizer 1
There are no transcripts associated with this gene.
- Gene
- SYNGR3
- Gene Name
- synaptogyrin 3
There are no transcripts associated with this gene.
- Gene
- TBL3
- Gene Name
- transducin beta like 3
There are no transcripts associated with this gene.
View Full Product Details