Product Details

SNP ID
rs11910678
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:41465059 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCGCCGCCACCATGGGCACTTTGC[C/T]TCAGCACATTCATTTATAGAACGTT
Phenotype
MIM: 147150 MIM: 602060
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MX1 PubMed Links

Gene Details

Gene
MX1
Gene Name
MX dynamin like GTPase 1
There are no transcripts associated with this gene.

Gene
TMPRSS2
Gene Name
transmembrane protease, serine 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135099.1 2696 UTR 3 NP_001128571.1
NM_005656.3 2696 UTR 3 NP_005647.3
XM_005261043.2 2696 UTR 3 XP_005261100.1
XM_011529731.2 2696 Intron XP_011528033.1
XM_011529732.2 2696 Intron XP_011528034.1
XM_011529733.1 2696 UTR 3 XP_011528035.1

View Full Product Details