Product Details

SNP ID
rs9916859
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:34574442 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGTTGGGCAAAAACCTGAATGAA[A/G]TAAAGGAGCAAGGGGTTAGGGGAAA
Phenotype
MIM: 616178
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C17orf102 PubMed Links

Gene Details

Gene
C17orf102
Gene Name
chromosome 17 open reading frame 102
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207454.2 3678 UTR 3 NP_997337.2
Gene
TMEM132E
Gene Name
transmembrane protein 132E
There are no transcripts associated with this gene.

View Full Product Details