Product Details

SNP ID
rs6526130
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:152832649 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGTCCCTCCATACCTTGCTTCCCA[C/G]TCCGTCAACCTGCAGATCGGAAGCC
Phenotype
MIM: 300006 MIM: 300275
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CETN2 PubMed Links

Gene Details

Gene
CETN2
Gene Name
centrin 2
There are no transcripts associated with this gene.

Gene
NSDHL
Gene Name
NAD(P) dependent steroid dehydrogenase-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129765.1 Intron NP_001123237.1
NM_015922.2 Intron NP_057006.1
XM_011531178.2 Intron XP_011529480.1
XM_017029564.1 Intron XP_016885053.1

View Full Product Details