Product Details
- SNP ID
-
rs35772513
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:62710461 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATCTTGGCAGAGCCCCAGCCTGTT[G/T]TCTTAATTGAGAGCTCAGGGTCTAA
- Phenotype
-
MIM: 613337
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC105371855
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111749478] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC105371855
- Gene Name
- uncharacterized LOC105371855
There are no transcripts associated with this gene.
- Gene
- MARCH10
- Gene Name
- membrane associated ring-CH-type finger 10
View Full Product Details