Product Details

SNP ID
rs2068657
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:62707582 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCGAGGCCTCAGAAGCATTGGCA[C/T]GTGTCCGCGAAGAGCCTAGCTGGAG
Phenotype
MIM: 613337
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105371855 PubMed Links

Gene Details

Gene
LOC105371855
Gene Name
uncharacterized LOC105371855
There are no transcripts associated with this gene.

Gene
MARCH10
Gene Name
membrane associated ring-CH-type finger 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100875.2 Intron NP_001094345.1
NM_001288779.1 Intron NP_001275708.1
NM_001288780.1 Intron NP_001275709.1
NM_152598.3 Intron NP_689811.2
XM_005257095.2 Intron XP_005257152.2
XM_005257096.2 Intron XP_005257153.2
XM_005257097.3 Intron XP_005257154.2
XM_005257098.2 Intron XP_005257155.2
XM_005257100.2 Intron XP_005257157.2
XM_005257102.2 Intron XP_005257159.2
XM_005257103.1 Intron XP_005257160.1
XM_005257106.4 Intron XP_005257163.2
XM_011524435.1 Intron XP_011522737.1
XM_011524436.1 Intron XP_011522738.1
XM_011524437.1 Intron XP_011522739.1

View Full Product Details