Product Details

SNP ID
rs11088489
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:39629352 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGTATTCTATACGTGCCATTTCAC[A/G]TGGTCTATTTCTCATTTTTCTTGTT
Phenotype
MIM: 604066
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
B3GALT5 PubMed Links

Gene Details

Gene
B3GALT5
Gene Name
beta-1,3-galactosyltransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278650.1 Intron NP_001265579.1
NM_006057.2 Intron NP_006048.1
NM_033170.2 Intron NP_149360.1
NM_033171.2 Intron NP_149361.1
NM_033172.2 Intron NP_149362.2
XM_017028227.1 Intron XP_016883716.1
XM_017028228.1 Intron XP_016883717.1
XM_017028229.1 Intron XP_016883718.1
XM_017028230.1 Intron XP_016883719.1
XM_017028231.1 Intron XP_016883720.1
XM_017028232.1 Intron XP_016883721.1
XM_017028233.1 Intron XP_016883722.1
XM_017028234.1 Intron XP_016883723.1
XM_017028235.1 Intron XP_016883724.1
XM_017028236.1 Intron XP_016883725.1
XM_017028237.1 Intron XP_016883726.1
XM_017028238.1 Intron XP_016883727.1
XM_017028239.1 Intron XP_016883728.1
XM_017028240.1 Intron XP_016883729.1
XM_017028241.1 Intron XP_016883730.1

View Full Product Details