Product Details

SNP ID
rs4829489
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:112633158 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCCAGATTGCTGGCTCAATAGTAC[A/C]GATGCATTTGTTTCTTTTCGTGATT
Phenotype
MIM: 300566
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LHFPL1 PubMed Links

Gene Details

Gene
LHFPL1
Gene Name
lipoma HMGIC fusion partner-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178175.3 Intron NP_835469.1
XM_011530943.2 Intron XP_011529245.1
XM_011530944.2 Intron XP_011529246.1
XM_011530946.2 Intron XP_011529248.1
XM_017029485.1 Intron XP_016884974.1

View Full Product Details