Product Details

SNP ID
rs11989719
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:89912090 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTACTTTTCTTATATCTTTGAGT[A/G]TATTTTATTTGGGGTAACATATTAG
Phenotype
MIM: 604598
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OSGIN2 PubMed Links

Gene Details

Gene
OSGIN2
Gene Name
oxidative stress induced growth inhibitor family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126111.2 Intron NP_001119583.1
NM_004337.2 Intron NP_004328.1
XM_011517287.2 Intron XP_011515589.1
XM_011517288.2 Intron XP_011515590.1

View Full Product Details