Product Details

SNP ID
rs6465352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:92145803 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAATTCTCTTATATGAGTATATTC[A/G]TTTTACCAGTGAAGAAACCAAGTCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYP51A1-AS1 PubMed Links

Gene Details

Gene
CYP51A1-AS1
Gene Name
CYP51A1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
LRRD1
Gene Name
leucine rich repeats and death domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161528.1 Intron NP_001155000.1

View Full Product Details