Product Details

SNP ID
rs2077080
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15644582 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGCGTCACCTTGCCTGTCACAAAG[C/G]TTAGAACAATGAGGACTTGTGGCTG
Phenotype
MIM: 601270
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CYP4F3 PubMed Links

Gene Details

Gene
CYP4F3
Gene Name
cytochrome P450 family 4 subfamily F member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000896.2 Intron NP_000887.2
NM_001199208.1 Intron NP_001186137.1
NM_001199209.1 Intron NP_001186138.1
XM_005259911.4 Intron XP_005259968.1
XM_011528014.2 Intron XP_011526316.1
XM_017026814.1 Intron XP_016882303.1
XM_017026815.1 Intron XP_016882304.1

View Full Product Details