Product Details

SNP ID
rs10241602
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:92609442 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATATACTTCAGACTTTTTTTTTT[A/T]AATTAGAGCTTCTTATGAATTTACA
Phenotype
MIM: 603368
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CDK6 PubMed Links

Gene Details

Gene
CDK6
Gene Name
cyclin dependent kinase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145306.1 7316 UTR 3 NP_001138778.1
NM_001259.6 7316 UTR 3 NP_001250.1
XM_006715835.2 7316 UTR 3 XP_006715898.1

View Full Product Details