Product Details

SNP ID
rs11979597
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:93907970 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTAAATAAAATGAACTTTTAGGAT[A/G]AAGAAAAGTTTTCTCTTCTTAAGTT
Phenotype
MIM: 189970
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNGT1 PubMed Links

Gene Details

Gene
GNGT1
Gene Name
G protein subunit gamma transducin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017012009.1 Intron XP_016867498.1

View Full Product Details