Product Details

SNP ID
rs1063192
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:22003368 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTTTCTTTAGTTTCCCTTAATATCA[A/G]GTTGTCATTAGGAAAGATTCCACAA
Phenotype
MIM: 600160 MIM: 600431 MIM: 613149
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDKN2A PubMed Links

Gene Details

Gene
CDKN2A
Gene Name
cyclin dependent kinase inhibitor 2A
There are no transcripts associated with this gene.

Gene
CDKN2B
Gene Name
cyclin dependent kinase inhibitor 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004936.3 3519 UTR 3 NP_004927.2
NM_078487.2 3519 UTR 3 NP_511042.1
Gene
CDKN2B-AS1
Gene Name
CDKN2B antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details