Product Details

SNP ID
rs36047398
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44833775 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTAGCTCTGTCATTTTCTATCTA[C/T]CAGTTCAATGTTTTTTGGCATAGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
H2AFV PubMed Links

Gene Details

Gene
H2AFV
Gene Name
H2A histone family member V
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012412.4 1154 UTR 3 NP_036544.1
NM_138635.3 1154 Intron NP_619541.1
NM_201436.2 1154 UTR 3 NP_958844.1
NM_201516.2 1154 UTR 3 NP_958924.1
NM_201517.2 1154 UTR 3 NP_958925.1

View Full Product Details