Product Details

SNP ID
rs9614037
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.22:29725296 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCTCGGCTGCTTGGGGAGTGGAAAT[A/G]AACGTGTCCCCTAACCAGGATTGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CABP7 PubMed Links

Gene Details

Gene
CABP7
Gene Name
calcium binding protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182527.2 Intron NP_872333.1
XM_006724161.3 Intron XP_006724224.1
Gene
ZMAT5
Gene Name
zinc finger matrin-type 5
There are no transcripts associated with this gene.

View Full Product Details