Product Details
- SNP ID
-
rs1045119
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:141661192 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGGTCACAGACATGGATGCACTGA[C/T]GGCTATTCCCATAACCGAGCCAGGC
- Phenotype
-
MIM: 610345
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AGK
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79301282] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AGK
- Gene Name
- acylglycerol kinase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018238.3 |
2832 |
Intron |
|
|
NP_060708.1 |
- Gene
- KIAA1147
- Gene Name
- KIAA1147
View Full Product Details