Product Details

SNP ID
rs6964456
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:141717046 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTACAGTTTTAAGTCAGTGAAATA[C/T]TAAATTGCCATTAAGATGTTTATAA
Phenotype
MIM: 614084
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WEE2 PubMed Links

Gene Details

Gene
WEE2
Gene Name
WEE1 homolog 2 (S. pombe)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105558.1 Intron NP_001099028.1
Gene
WEE2-AS1
Gene Name
WEE2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details