Product Details

SNP ID
rs11974752
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:21545392 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATGCTTACTGATGCTAATATTCA[A/G]CAGGTGGCAATTGCAGGGATCATTC
Phenotype
MIM: 603339
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAH11 PubMed Links

Gene Details

Gene
DNAH11
Gene Name
dynein axonemal heavy chain 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277115.1 Intron NP_001264044.1

View Full Product Details