Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001076674.1 | Intron | NP_001070142.1 | |||
NM_024107.2 | Intron | NP_077012.2 | |||
NM_177441.2 | Intron | NP_803190.2 | |||
XM_011525198.2 | Intron | XP_011523500.1 | |||
XM_011525200.2 | Intron | XP_011523502.1 | |||
XM_011525201.2 | Intron | XP_011523503.1 | |||
XM_011525203.2 | Intron | XP_011523505.1 | |||
XM_011525204.2 | Intron | XP_011523506.1 | |||
XM_011525205.2 | Intron | XP_011523507.1 | |||
XM_011525206.2 | Intron | XP_011523508.1 | |||
XM_011525207.2 | Intron | XP_011523509.1 | |||
XM_011525208.2 | Intron | XP_011523510.1 | |||
XM_011525209.2 | Intron | XP_011523511.1 | |||
XM_011525210.2 | Intron | XP_011523512.1 | |||
XM_011525211.2 | Intron | XP_011523513.1 | |||
XM_017025047.1 | Intron | XP_016880536.1 | |||
XM_017025048.1 | Intron | XP_016880537.1 | |||
XM_017025049.1 | Intron | XP_016880538.1 | |||
XM_017025050.1 | Intron | XP_016880539.1 | |||
XM_017025051.1 | Intron | XP_016880540.1 | |||
XM_017025052.1 | Intron | XP_016880541.1 | |||
XM_017025053.1 | Intron | XP_016880542.1 | |||
XM_017025054.1 | Intron | XP_016880543.1 | |||
XM_017025055.1 | Intron | XP_016880544.1 | |||
XM_017025056.1 | Intron | XP_016880545.1 |