Product Details
- SNP ID
-
rs757556
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:48108710 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CAATTTATATGAAGCTTCAGAAACA[C/T]ATCTATGTATTTCGTCTTAACTTAA
- Phenotype
-
MIM: 610681
MIM: 612157
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PFKM
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17614932] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PFKM
- Gene Name
- phosphofructokinase, muscle
- Gene
- SENP1
- Gene Name
- SUMO1/sentrin specific peptidase 1
There are no transcripts associated with this gene.
View Full Product Details