Product Details

SNP ID
rs5752976
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.22:29885973 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GGCTTTGGTAAATATCCTGTCAAAT[C/G]ATAGAGTAGTATTTTTAGAGTTGCA
Phenotype
MIM: 603558
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MTMR3 PubMed Links

Gene Details

Gene
MTMR3
Gene Name
myotubularin related protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021090.3 Intron NP_066576.1
NM_153050.2 Intron NP_694690.1
NM_153051.2 Intron NP_694691.1
XM_005261803.2 Intron XP_005261860.1
XM_005261804.3 Intron XP_005261861.1
XM_005261805.2 Intron XP_005261862.1
XM_005261806.2 Intron XP_005261863.1
XM_005261808.2 Intron XP_005261865.1
XM_017029028.1 Intron XP_016884517.1
XM_017029029.1 Intron XP_016884518.1
XM_017029030.1 Intron XP_016884519.1
XM_017029031.1 Intron XP_016884520.1
XM_017029032.1 Intron XP_016884521.1
XM_017029033.1 Intron XP_016884522.1
XM_017029034.1 Intron XP_016884523.1

View Full Product Details