Product Details

SNP ID
rs11743810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:138466715 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTCGTCCTCGTCCTCCAGTGATTG[C/T]TTTCCAGTAACCAGGCCTCCCGCTT
Phenotype
MIM: 128990
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EGR1 PubMed Links

Gene Details

Gene
EGR1
Gene Name
early growth response 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001964.2 Intron NP_001955.1

View Full Product Details