Product Details

SNP ID
rs11089450
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30450657 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACAAGTAAGACCTCTCCGTTTCAA[G/T]TTATGGGACCCTGGGCAAATTCCTT
Phenotype
MIM: 612824
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SEC14L3 PubMed Links

Gene Details

Gene
SEC14L3
Gene Name
SEC14 like lipid binding 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257378.1 Intron NP_001244307.1
NM_001257379.1 Intron NP_001244308.1
NM_001257382.1 Intron NP_001244311.1
NM_174975.4 Intron NP_777635.1
XM_011530128.2 Intron XP_011528430.1

View Full Product Details