Product Details

SNP ID
rs13022757
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:101997084 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGGGACATTGTGTCCCCAGGGAGT[C/G]GTGGGCAGCTGATCCACACGTGCTG
Phenotype
MIM: 147811
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IL1R2 PubMed Links

Gene Details

Gene
IL1R2
Gene Name
interleukin 1 receptor type 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261419.1 235 Intron NP_001248348.1
NM_004633.3 235 Intron NP_004624.1
XM_006712734.3 235 Intron XP_006712797.1
XM_006712736.3 235 Intron XP_006712799.1
XM_011511801.2 235 Intron XP_011510103.1
XM_011511803.2 235 UTR 5 XP_011510105.1
XM_011511804.2 235 Intron XP_011510106.1
XM_011511805.2 235 Intron XP_011510107.1
XM_011511807.1 235 Intron XP_011510109.1
XM_011511808.2 235 Intron XP_011510110.1
XM_017004889.1 235 Intron XP_016860378.1

View Full Product Details