Product Details

SNP ID
rs1183739
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74529421 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGGAAGACGTGGATCCCGAGAAA[C/G]AGGCGCAGGACGAGGAGGCAGAACC
Phenotype
MIM: 602434 MIM: 606441 MIM: 607163
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AUP1 PubMed Links

Gene Details

Gene
AUP1
Gene Name
ancient ubiquitous protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181575.4 17 Silent Mutation CTC,CTG L43L NP_853553.1
XM_005264392.3 17 Missense Mutation TCT,TGT S74C XP_005264449.1
Gene
DQX1
Gene Name
DEAQ-box RNA dependent ATPase 1
There are no transcripts associated with this gene.

Gene
HTRA2
Gene Name
HtrA serine peptidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321727.1 17 UTR 5 NP_001308656.1
NM_001321728.1 17 UTR 5 NP_001308657.1
NM_013247.4 17 UTR 5 NP_037379.1
NM_145074.2 17 UTR 5 NP_659540.1
Gene
LOXL3
Gene Name
lysyl oxidase like 3
There are no transcripts associated with this gene.

View Full Product Details