Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001256007.2 | Intron | NP_001242936.1 | |||
NM_001256008.2 | Intron | NP_001242937.1 | |||
NM_001256009.2 | Intron | NP_001242938.1 | |||
NM_001256010.2 | Intron | NP_001242939.1 | |||
NM_001256011.2 | Intron | NP_001242940.1 | |||
NM_015723.4 | Intron | NP_056538.1 | |||
XM_005250396.4 | Intron | XP_005250453.1 | |||
XM_011516274.2 | Intron | XP_011514576.1 | |||
XM_011516275.2 | Intron | XP_011514577.1 | |||
XM_017012261.1 | Intron | XP_016867750.1 |