Product Details

SNP ID
rs12410155
Assay Type
Functionally tested
NCBI dbSNP Submissions
50
Location
Chr.1:43722794 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGATTGAGATAGTAAATGCTGGAGG[A/C]AGAGTAATTCCTCGGGGTGAAACAT
Phenotype
MIM: 606494
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929592 PubMed Links

Gene Details

Gene
LOC101929592
Gene Name
uncharacterized LOC101929592
There are no transcripts associated with this gene.

Gene
ST3GAL3
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270459.1 Intron NP_001257388.1
NM_001270460.1 Intron NP_001257389.1
NM_001270461.1 Intron NP_001257390.1
NM_001270462.1 Intron NP_001257391.1
NM_001270463.1 Intron NP_001257392.1
NM_001270464.1 Intron NP_001257393.1
NM_001270465.1 Intron NP_001257394.1
NM_001270466.1 Intron NP_001257395.1
NM_006279.3 Intron NP_006270.1
NM_174963.3 Intron NP_777623.2
NM_174964.2 Intron NP_777624.1
NM_174965.2 Intron NP_777625.1
NM_174966.2 Intron NP_777626.1
NM_174967.2 Intron NP_777627.1
NM_174968.3 Intron NP_777628.2
NM_174969.2 Intron NP_777629.1
NM_174970.2 Intron NP_777630.1
NM_174971.3 Intron NP_777631.2
XM_006710825.3 Intron XP_006710888.1
XM_006710827.3 Intron XP_006710890.1
XM_006710828.3 Intron XP_006710891.1
XM_011541973.2 Intron XP_011540275.1
XM_011541974.2 Intron XP_011540276.1
XM_011541976.2 Intron XP_011540278.1
XM_011541977.2 Intron XP_011540279.1
XM_011541980.2 Intron XP_011540282.1
XM_011541981.2 Intron XP_011540283.1
XM_011541983.2 Intron XP_011540285.1
XM_011541984.2 Intron XP_011540286.1
XM_011541985.2 Intron XP_011540287.1
XM_011541986.2 Intron XP_011540288.1
XM_011541987.2 Intron XP_011540289.1
XM_011541988.2 Intron XP_011540290.1
XM_017002109.1 Intron XP_016857598.1
XM_017002110.1 Intron XP_016857599.1
XM_017002111.1 Intron XP_016857600.1
XM_017002112.1 Intron XP_016857601.1
XM_017002113.1 Intron XP_016857602.1
XM_017002114.1 Intron XP_016857603.1
XM_017002115.1 Intron XP_016857604.1
XM_017002116.1 Intron XP_016857605.1
XM_017002117.1 Intron XP_016857606.1
XM_017002118.1 Intron XP_016857607.1
XM_017002119.1 Intron XP_016857608.1
XM_017002120.1 Intron XP_016857609.1
XM_017002121.1 Intron XP_016857610.1
XM_017002122.1 Intron XP_016857611.1

View Full Product Details