Product Details

SNP ID
rs4717907
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:74672169 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AATGAGTTCTTTGGCTCTATTTCTT[A/G]TCTGCTGAAGTGTATATGTTAATAT
Phenotype
MIM: 601679
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GTF2I PubMed Links

Gene Details

Gene
GTF2I
Gene Name
general transcription factor IIi
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163636.2 Intron NP_001157108.1
NM_001280800.1 Intron NP_001267729.1
NM_001518.4 Intron NP_001509.3
NM_032999.3 Intron NP_127492.1
NM_033000.3 Intron NP_127493.1
NM_033001.3 Intron NP_127494.1

View Full Product Details