Product Details

SNP ID
rs8012627
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:73487792 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCTTGAGATGAAGGAGATGGTAGCG[A/C]CTTTGGAGAGGAGATGGAATGTGAA
Phenotype
MIM: 614313 MIM: 611919
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ACOT1 PubMed Links

Gene Details

Gene
ACOT1
Gene Name
acyl-CoA thioesterase 1
There are no transcripts associated with this gene.

Gene
C14orf169
Gene Name
chromosome 14 open reading frame 169
There are no transcripts associated with this gene.

Gene
HEATR4
Gene Name
HEAT repeat containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001220484.1 Intron NP_001207413.1
NM_203309.2 Intron NP_976054.2
XM_006720143.2 Intron XP_006720206.1
XM_011536760.2 Intron XP_011535062.1
XM_011536761.2 Intron XP_011535063.1
XM_017021289.1 Intron XP_016876778.1

View Full Product Details