Product Details

SNP ID
rs2240389
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:101613553 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTTGCCGCACACATCTGGGGGAAA[C/T]GCTGCAAACATCTGCTTGACCTGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYL10 PubMed Links

Gene Details

Gene
MYL10
Gene Name
myosin light chain 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138403.4 501 Silent Mutation GCA,GCG A201A NP_612412.2
XM_017012793.1 501 Silent Mutation GCA,GCG A122A XP_016868282.1

View Full Product Details