Product Details

SNP ID
rs12170331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:17117519 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGCGGGGGCTTGGCCTGGGTGCT[A/G]AGTTAACGGCCAGTGACTTGGCACT
Phenotype
MIM: 605461
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CECR6 PubMed Links

Gene Details

Gene
CECR6
Gene Name
cat eye syndrome chromosome region, candidate 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163079.1 2741 UTR 3 NP_001156551.1
NM_031890.3 2741 UTR 3 NP_114096.1
XM_011546124.2 2741 Intron XP_011544426.1
XM_011546125.2 2741 UTR 3 XP_011544427.1
Gene
IL17RA
Gene Name
interleukin 17 receptor A
There are no transcripts associated with this gene.

Gene
LOC100996342
Gene Name
uncharacterized LOC100996342
There are no transcripts associated with this gene.

View Full Product Details