Product Details
- SNP ID
-
rs2098029
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:73960702 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGGAAGACCGTAAATGACAATCTC[C/T]GTACAGTGATAGAAGTATGCATAGG
- Phenotype
-
MIM: 614647
MIM: 603162
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
COQ6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115579294] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- COQ6
- Gene Name
- coenzyme Q6, monooxygenase
- Gene
- ENTPD5
- Gene Name
- ectonucleoside triphosphate diphosphohydrolase 5
View Full Product Details