Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_182476.2 | 173 | Intron | NP_872282.1 | ||
NM_182480.2 | 173 | Intron | NP_872286.2 | ||
XM_006720156.1 | 173 | Intron | XP_006720219.1 | ||
XM_011536807.1 | 173 | Intron | XP_011535109.1 | ||
XM_011536808.1 | 173 | Intron | XP_011535110.1 | ||
XM_011536809.2 | 173 | Intron | XP_011535111.1 | ||
XM_011536810.2 | 173 | Intron | XP_011535112.1 | ||
XM_017021351.1 | 173 | Intron | XP_016876840.1 | ||
XM_017021352.1 | 173 | Intron | XP_016876841.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_152445.2 | 173 | UTR 5 | NP_689658.2 | ||
XM_011536475.1 | 173 | Intron | XP_011534777.1 |