Product Details

SNP ID
rs10133554
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:74308190 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAACTTTGGGAGGCCAAGGCCAGA[A/G]GTTTGCTTGAGCCCAGGAGTTCAAG
Phenotype
MIM: 603214
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCD4 PubMed Links

Gene Details

Gene
ABCD4
Gene Name
ATP binding cassette subfamily D member 4
There are no transcripts associated with this gene.

Gene
VRTN
Gene Name
vertebrae development associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018228.2 Intron NP_060698.2
XM_011536911.1 Intron XP_011535213.1

View Full Product Details