Product Details

SNP ID
rs7805578
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:156639037 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTAATCATACGTCCCATCCCAGGA[C/T]TGAAGAAGTACCAGGGCTGTTGCTG
Phenotype
MIM: 610242 MIM: 610241
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C7orf13 PubMed Links

Gene Details

Gene
C7orf13
Gene Name
chromosome 7 open reading frame 13
There are no transcripts associated with this gene.

Gene
RNF32
Gene Name
ring finger protein 32
There are no transcripts associated with this gene.

View Full Product Details