Product Details

SNP ID
rs34927032
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:30424892 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCAGCCTTCTAGAGGAGGGTAGC[C/T]TAGCAGAGGAGAAGCCCTGAGTGGA
Phenotype
MIM: 605980
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
NOD1 PubMed Links

Gene Details

Gene
NOD1
Gene Name
nucleotide binding oligomerization domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006092.2 4045 UTR 3 NP_006083.1
XM_005249568.1 4045 UTR 3 XP_005249625.1
XM_005249572.1 4045 UTR 3 XP_005249629.1
XM_005249576.1 4045 UTR 3 XP_005249633.1
XM_006715633.2 4045 UTR 3 XP_006715696.1
XM_011515079.1 4045 UTR 3 XP_011513381.1
XM_011515080.2 4045 UTR 3 XP_011513382.1
XM_011515081.2 4045 UTR 3 XP_011513383.1
XM_011515083.1 4045 UTR 3 XP_011513385.1
XM_011515084.1 4045 UTR 3 XP_011513386.1
XM_011515085.1 4045 UTR 3 XP_011513387.1
XM_011515087.1 4045 Intron XP_011513389.1
XM_011515088.2 4045 Intron XP_011513390.1
XM_017011674.1 4045 Intron XP_016867163.1
XM_017011675.1 4045 UTR 3 XP_016867164.1

View Full Product Details