Product Details

SNP ID
rs10033825
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:10495168 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTAGTTTATCATGTAAATATCTA[C/T]GGTGGTTTTTAGAAACTTATAAACA
Phenotype
MIM: 611434
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLNK PubMed Links

Gene Details

Gene
CLNK
Gene Name
cytokine dependent hematopoietic cell linker
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052964.2 Intron NP_443196.2
XM_011513775.2 Intron XP_011512077.1
XM_017007684.1 Intron XP_016863173.1
XM_017007685.1 Intron XP_016863174.1

View Full Product Details