Product Details

SNP ID
rs2429369
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:58424059 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTAAAAGACAGACTAGTCACCAGGG[A/G]TAGGTGCTTACTCATCTGGTGAACT
Phenotype
MIM: 612482
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSF5 PubMed Links

Gene Details

Gene
HSF5
Gene Name
heat shock transcription factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080439.2 Intron NP_001073908.2
XM_011524283.1 Intron XP_011522585.1
Gene
RNF43
Gene Name
ring finger protein 43
There are no transcripts associated with this gene.

View Full Product Details