Product Details

SNP ID
rs1057947
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:8113349 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCCATAGTAAGGTTTGTGCCTTGGA[C/G]AGAAGCCCAGCTCCTCCACCAGTGT
Phenotype
MIM: 147625
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ICA1 PubMed Links

Gene Details

Gene
ICA1
Gene Name
islet cell autoantigen 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136020.2 2093 UTR 3 NP_001129492.1
NM_001276478.1 2093 UTR 3 NP_001263407.1
NM_004968.3 2093 UTR 3 NP_004959.2
NM_022307.2 2093 UTR 3 NP_071682.1
XM_005249735.3 2093 Intron XP_005249792.1
XM_011515345.1 2093 Intron XP_011513647.1
XM_011515346.1 2093 Intron XP_011513648.1
XM_011515347.2 2093 Intron XP_011513649.1
XM_011515348.1 2093 Intron XP_011513650.1
XM_011515349.1 2093 Intron XP_011513651.1
XM_011515350.1 2093 Intron XP_011513652.1
XM_011515351.1 2093 Intron XP_011513653.1
XM_011515352.1 2093 Intron XP_011513654.1
XM_011515353.2 2093 Intron XP_011513655.1
XM_011515354.1 2093 Intron XP_011513656.1
XM_011515355.2 2093 Intron XP_011513657.1
XM_011515356.2 2093 Intron XP_011513658.1
XM_011515357.2 2093 Intron XP_011513659.1
XM_017012116.1 2093 Intron XP_016867605.1
XM_017012117.1 2093 Intron XP_016867606.1
XM_017012118.1 2093 Intron XP_016867607.1
XM_017012119.1 2093 Intron XP_016867608.1
XM_017012120.1 2093 Intron XP_016867609.1
XM_017012121.1 2093 Intron XP_016867610.1
XM_017012122.1 2093 Intron XP_016867611.1
XM_017012123.1 2093 Intron XP_016867612.1
XM_017012124.1 2093 Intron XP_016867613.1
XM_017012125.1 2093 Intron XP_016867614.1

View Full Product Details