Product Details

SNP ID
rs11836052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:3615278 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCGGGCCCCGGTGGGTGGCATC[C/T]GTGCAGGAGCCGTGTGCAGGATCAG
Phenotype
MIM: 614178
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRACR2A PubMed Links

Gene Details

Gene
CRACR2A
Gene Name
calcium release activated channel regulator 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144958.1 1693 Intron NP_001138430.1
NM_032680.3 1693 Intron NP_116069.1
XM_006719021.3 1693 UTR 3 XP_006719084.1
XM_011521034.2 1693 UTR 3 XP_011519336.1
XM_011521036.2 1693 UTR 3 XP_011519338.1
XM_011521037.2 1693 UTR 3 XP_011519339.1
XM_011521038.2 1693 UTR 3 XP_011519340.1

View Full Product Details