Product Details

SNP ID
rs2240920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52796993 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACGTTGAGGCTGAGGCAAGTGAGG[C/T]TCAGAATGGTTAGGGATCTCCCTCG
Phenotype
MIM: 147270 MIM: 146650
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ITIH1 PubMed Links

Gene Details

Gene
ITIH1
Gene Name
inter-alpha-trypsin inhibitor heavy chain 1
There are no transcripts associated with this gene.

Gene
ITIH3
Gene Name
inter-alpha-trypsin inhibitor heavy chain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002217.3 Intron NP_002208.3
XM_005265105.4 Intron XP_005265162.1
XM_017006356.1 Intron XP_016861845.1

View Full Product Details