Product Details

SNP ID
rs3755798
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52707144 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCAGGAGTTTTCATGGTGCTCAC[A/G]TTGTCCCTTCACCTTTTATTTATTT
Phenotype
MIM: 601959 MIM: 610358
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLT8D1 PubMed Links

Gene Details

Gene
GLT8D1
Gene Name
glycosyltransferase 8 domain containing 1
There are no transcripts associated with this gene.

Gene
NEK4
Gene Name
NIMA related kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193533.1 Intron NP_001180462.1
NM_003157.4 Intron NP_003148.2
XM_006713310.2 Intron XP_006713373.1
XM_011534039.2 Intron XP_011532341.1
XM_011534040.2 Intron XP_011532342.1
XM_017007085.1 Intron XP_016862574.1
XM_017007086.1 Intron XP_016862575.1
Gene
SPCS1
Gene Name
signal peptidase complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014041.3 Intron NP_054760.3

View Full Product Details