Product Details

SNP ID
rs880108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43610988 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGCCTTGGTATGGGCTGTTCCCCA[A/C]TCTATCCACCTGGTGACCCCCTACT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SRRM5 PubMed Links

Gene Details

Gene
SRRM5
Gene Name
serine/arginine repetitive matrix 5
There are no transcripts associated with this gene.

Gene
ZNF428
Gene Name
zinc finger protein 428
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182498.3 Intron NP_872304.2

View Full Product Details