Product Details

SNP ID
rs12972381
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56369375 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGAAATTTTGGGGAATGGGAATGT[C/G]ATTGATATCTTAAGCATGAAAGTCC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF542P PubMed Links

Gene Details

Gene
ZNF542P
Gene Name
zinc finger protein 542, pseudogene
There are no transcripts associated with this gene.

Gene
ZSCAN5A
Gene Name
zinc finger and SCAN domain containing 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322061.1 Intron NP_001308990.1
NM_001322062.1 Intron NP_001308991.1
NM_001322064.1 Intron NP_001308993.1
NM_001322065.1 Intron NP_001308994.1
NM_001322066.1 Intron NP_001308995.1
NM_001322067.1 Intron NP_001308996.1
NM_001322068.1 Intron NP_001308997.1
NM_001322069.1 Intron NP_001308998.1
NM_001322070.1 Intron NP_001308999.1
NM_001322072.1 Intron NP_001309001.1
NM_001322073.1 Intron NP_001309002.1
NM_001322074.1 Intron NP_001309003.1
NM_001322075.1 Intron NP_001309004.1
NM_001322076.1 Intron NP_001309005.1
NM_001322077.1 Intron NP_001309006.1
NM_001322078.1 Intron NP_001309007.1
NM_024303.2 Intron NP_077279.1
XM_005259254.1 Intron XP_005259311.1
XM_006723377.3 Intron XP_006723440.1
XM_011527308.1 Intron XP_011525610.1
XM_017027298.1 Intron XP_016882787.1
XM_017027299.1 Intron XP_016882788.1

View Full Product Details